ENA370563586 Enamine

N-(2-chloroethyl)cyclopentanecarboxamide

  • MDL number MFCD14613677
  • Cas Number : 1250039-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-106313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106313
Availability details for
Quantity
  • Availability
  • Add to Cart