ENA370563587 Enamine

4-methyl-2-(methylsulfanyl)benzoic acid

  • MDL number MFCD16853043
  • Cas Number : 24852-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106315

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106315
Availability details for
Quantity
  • Availability
  • Add to Cart