ENA370563588 Enamine

4-(2,5-dichlorothiophen-3-yl)-4-oxobutanoic acid

  • MDL number MFCD11642116
  • Cas Number : 19945-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101435
Availability details for
Quantity
  • Availability
  • Add to Cart