ENA370563589 Enamine

N-cyclopentyl-3-fluoroaniline

  • MDL number MFCD11144485
  • Cas Number : 950494-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-106319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106319
Availability details for
Quantity
  • Availability
  • Add to Cart