ENA370563590 Enamine

2-methyl-6-(1H-pyrrol-1-ylmethyl)pyridine

  • MDL number MFCD16704784
  • Cas Number : 1251105-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106320
Availability details for
Quantity
  • Availability
  • Add to Cart