ENA370563591 Enamine

6,7,8,9-tetrahydrospiro[benzo[7]annulene-5,3'-piperidine]

  • MDL number MFCD19680400
  • Cas Number : 1342594-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106321
Availability details for
Quantity
  • Availability
  • Add to Cart