ENA370563592 Enamine

(2-methoxy-4-methylphenyl)methanamine

  • MDL number MFCD16704878
  • Cas Number : 851670-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106322

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106322
Availability details for
Quantity
  • Availability
  • Add to Cart