ENA370563594 Enamine

5-(1-chloroethyl)-3-[(4-fluorophenyl)methyl]-1,2,4-oxadiazole

  • MDL number MFCD11205895
  • Cas Number : 1094291-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104279

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104279
Availability details for
Quantity
  • Availability
  • Add to Cart