ENA370563596 Enamine

2-cyano-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD16610733
  • Cas Number : 779-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106349

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106349
Availability details for
Quantity
  • Availability
  • Add to Cart