ENA370563597 Enamine

3-{[4-(1H-1,2,3,4-tetrazol-5-yl)phenyl]formamido}propanoic acid

  • MDL number MFCD12544347
  • Cas Number : 1099107-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104280
Availability details for
Quantity
  • Availability
  • Add to Cart