ENA370563605 Enamine

propan-2-yl 2-[(4-methoxyphenyl)methoxy]acetate

  • MDL number MFCD16214926
  • Cas Number : 1274982-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106389
Availability details for
Quantity
  • Availability
  • Add to Cart