ENA370563610 Enamine

2-chloro-4-(1H-pyrazol-1-yl)pyridine

  • MDL number MFCD14702706
  • Cas Number : 1209459-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106395

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106395
Availability details for
Quantity
  • Availability
  • Add to Cart