ENA370563614 Enamine

N-methyl-7-nitro-2,1,3-benzoxadiazol-4-amine

  • MDL number MFCD00576809
  • Cas Number : 18378-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258-260
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106403
Availability details for
Quantity
  • Availability
  • Add to Cart