ENA370563615 Enamine

(2-fluoro-4-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD04116339
  • Cas Number : 937783-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217...219
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106409

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-106409
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart