ENA370563616 Enamine

2-[4-(4-methylphenyl)-2-phenyl-1,3-thiazol-5-yl]acetic acid

  • MDL number MFCD09711925
  • Cas Number : 23821-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-169
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106414
Availability details for
Quantity
  • Availability
  • Add to Cart