ENA370563633 Enamine

2-chloro-N-(2,6-diethylphenyl)propanamide

  • MDL number MFCD09946634
  • Cas Number : 1016871-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27940
Availability details for
Quantity
  • Availability
  • Add to Cart