ENA370563635 Enamine

2-bromo-N-cyclohexyl-N,3-dimethylbutanamide

  • MDL number MFCD09931003
  • Cas Number : 1016508-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29144
Availability details for
Quantity
  • Availability
  • Add to Cart