ENA370563637 Enamine

N-(2-methylpropyl)cyclopentanamine

  • MDL number MFCD11139813
  • Cas Number : 42870-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-32550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32550
Availability details for
Quantity
  • Availability
  • Add to Cart