ENA370563658 Enamine

(2,4-dichloro-6-methylpyridin-3-yl)methanol

  • MDL number MFCD08236805
  • Cas Number : 374800-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54237

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54237
Availability details for
Quantity
  • Availability
  • Add to Cart