ENA370563660 Enamine

ethyl 1-oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate

  • MDL number MFCD00973033
  • Cas Number : 6742-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56328

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56328
Availability details for
Quantity
  • Availability
  • Add to Cart