ENA370563661 Enamine

2-hydrazinyl-4-methylpyridine

  • MDL number MFCD09863246
  • Cas Number : 4931-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-75
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56339
Availability details for
Quantity
  • Availability
  • Add to Cart