ENA370563662 Enamine

3-(benzyloxy)-2-methylpropan-1-amine

  • MDL number MFCD11653689
  • Cas Number : 114377-41-2
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58780
Availability details for
Quantity
  • Availability
  • Add to Cart