ENA370563668 Enamine

1,3-dimethyl-5-(2,2,2-trifluoroethoxy)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11191307
  • Cas Number : 126764-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63796

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63796
Availability details for
Quantity
  • Availability
  • Add to Cart