ENA370563678 Enamine

2-[(2-methylpropyl)amino]acetic acid

  • MDL number MFCD11185547
  • Cas Number : 3182-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67561
Availability details for
Quantity
  • Availability
  • Add to Cart