ENA370563680 Enamine

tert-butyl N-[4-(2-aminoethyl)phenyl]carbamate

  • MDL number MFCD09701235
  • Cas Number : 457631-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69394
Availability details for
Quantity
  • Availability
  • Add to Cart