ENA370563698 Enamine

2-(2,3-dihydro-1H-indol-1-yl)-N'-hydroxyethanimidamide

  • MDL number MFCD09942601
  • Cas Number : 1016831-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79662
Availability details for
Quantity
  • Availability
  • Add to Cart