ENA370563700 Enamine

2-{[(9H-fluoren-9-ylmethoxy)carbonyl](2-methylpropyl)amino}acetic acid

  • MDL number MFCD18351999
  • Cas Number : 141743-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81384

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81384
Availability details for
Quantity
  • Availability
  • Add to Cart