ENA370563715 Enamine

1-ethyl-3-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD22369997
  • Cas Number : 1384427-70-6
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-90906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90906
Availability details for
Quantity
  • Availability
  • Add to Cart