ENA370563739 Enamine

4-chloro-6-hydrazinyl-2-(methylsulfanyl)pyrimidine

  • MDL number MFCD00974394
  • Cas Number : 89280-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93160

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93160
Availability details for
Quantity
  • Availability
  • Add to Cart