ENA370563750 Enamine

4-nitro-1H-pyrazol-5-amine

  • MDL number MFCD00475931
  • Cas Number : 16115-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237...239
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85837

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-85837
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart