ENA370563763 Enamine

4-methoxy-2-methylidene-4-oxobutanoic acid

  • MDL number MFCD00021713
  • Cas Number : 7338-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97200

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-97200
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart