ENA370563772 Enamine

(1R)-1-(oxolan-3-yl)ethan-1-ol

  • MDL number MFCD21188302
  • Cas Number : 1372908-76-3
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-tetrahydro-3-furanylethanol
Inchi Code 1S/C6H12O2/c1-5(7)6-2-3-8-4-6/h5-7H,2-4H2,1H3/t5-,6?/m1/s1
Inchi Key ARDSHQRPKXPELC-LWOQYNTDSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-98715

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-98715
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart