ENA370563779 Enamine

ethyl 5-methyl-3-oxohexanoate

  • MDL number MFCD00086868
  • Cas Number : 34036-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99421

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99421
Availability details for
Quantity
  • Availability
  • Add to Cart