ENA370563784 Enamine

[1-(3,5-dimethoxyphenyl)pyrrolidin-3-yl]methanamine

  • MDL number MFCD09944506
  • Cas Number : 1016820-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99819
Availability details for
Quantity
  • Availability
  • Add to Cart