ENA371177384 Enamine

3-(chloromethyl)-7-phenyl-3H,4H,7H-pyrazolo[3,4-d][1,2,3]triazin-4-one

  • MDL number MFCD22375216
  • Cas Number : 1394042-50-2
  • Molecular Weight : 261.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101279
Availability details for
Quantity
  • Availability
  • Add to Cart