ENA371177393 Enamine

2-(1-chloroethyl)-6-methylpyrimidin-4-ol

  • MDL number MFCD22375235
  • Cas Number : 1394040-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-105843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105843
Availability details for
Quantity
  • Availability
  • Add to Cart