ENA371177394 Enamine

5-(chlorosulfonyl)-2-nitrobenzoic acid

  • MDL number MFCD22375236
  • Cas Number : 98555-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105920
Availability details for
Quantity
  • Availability
  • Add to Cart