ENA371177395 Enamine

2-bromo-3-[(1,3-thiazol-2-yl)imino]propanal

  • Cas Number : 1394045-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106076
Availability details for
Quantity
  • Availability
  • Add to Cart