ENA371177396 Enamine

1-(2,4-dimethylphenyl)-5-(4-fluorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD22375239
  • Cas Number : 1394041-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106196
Availability details for
Quantity
  • Availability
  • Add to Cart