ENA371177411 Enamine

1-[(3-chlorophenyl)methyl]piperazin-2-one hydrochloride

  • MDL number MFCD22375267
  • Cas Number : 1394041-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106640
Availability details for
Quantity
  • Availability
  • Add to Cart