ENA371177412 Enamine

[3-(4-methylphenyl)phenyl]methanamine hydrochloride

  • MDL number MFCD22375273
  • Cas Number : 1394042-69-3
  • Molecular Weight : 233.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106680
Availability details for
Quantity
  • Availability
  • Add to Cart