ENA371177414 Enamine

methyl 3-(2-methoxyethyl)pyrrolidine-3-carboxylate

  • MDL number MFCD22375275
  • Cas Number : 1384510-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106684
Availability details for
Quantity
  • Availability
  • Add to Cart