ENA371177419 Enamine

2-(tert-butoxy)-6-(2-isocyanatoethyl)pyridine

  • MDL number MFCD22375287
  • Cas Number : 1394040-23-3
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-tert-butoxy-6-(2-isocyanatoethyl)pyridine
Inchi Code 1S/C12H16N2O2/c1-12(2,3)16-11-6-4-5-10(14-11)7-8-13-9-15/h4-6H,7-8H2,1-3H3
Inchi Key AWBBZPMKCVJVLL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-106769

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P312,P302+P352,P304+P340,P304+P341,P305+P351+P338,P312,P330,P332+P313,P337+P313,P342+P311,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-106769
pictograms GHS07,GHS08
hstatements H302,H312,H315,H319,H332,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart