ENA371177420 Enamine

N-[4-(aminomethyl)phenyl]-4-fluorobenzamide hydrochloride

  • MDL number MFCD22375289
  • Cas Number : 1394041-06-5
  • Molecular Weight : 280.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[4-(aminomethyl)phenyl]-4-fluorobenzamide hydrochloride
Inchi Code 1S/C14H13FN2O.ClH/c15-12-5-3-11(4-6-12)14(18)17-13-7-1-10(9-16)2-8-13;/h1-8H,9,16H2,(H,17,18);1H
Inchi Key XESGSIPBKRPYAA-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-106776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106776
Availability details for
Quantity
  • Availability
  • Add to Cart