ENA371177426 Enamine

(3-fluoro-5-methoxyphenyl)hydrazine hydrochloride

  • MDL number MFCD22375302
  • Cas Number : 1394041-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106897
Availability details for
Quantity
  • Availability
  • Add to Cart