ENA371177427 Enamine

7-bromo-5-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD22375305
  • Cas Number : 1394041-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106900
Availability details for
Quantity
  • Availability
  • Add to Cart