ENA371177430 Enamine

1-[2-(pyrrolidin-3-yloxy)ethyl]piperidine dihydrochloride

  • MDL number MFCD22375309
  • Cas Number : 1394042-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106987
Availability details for
Quantity
  • Availability
  • Add to Cart