ENA371177435 Enamine

2-cyano-3-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD22375317
  • Cas Number : 1394040-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107052
Availability details for
Quantity
  • Availability
  • Add to Cart