ENA371177446 Enamine

3-ethoxy-4-methylpentanoic acid

  • MDL number MFCD22375339
  • Cas Number : 1394040-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107274
Availability details for
Quantity
  • Availability
  • Add to Cart