ENA371177448 Enamine

(4-bromobutyl)[(4-fluorophenyl)methyl]methylamine hydrobromide

  • MDL number MFCD22375343
  • Cas Number : 1394041-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107304
Availability details for
Quantity
  • Availability
  • Add to Cart